Would you like to log in again?
COLUMN DC C8 4.6X150 3U
Supplier: Agela Technologies
Column Durashell C8 4.6X150mm 3um^ Reverse Phase Columns for Small Molecules; Wide pH range (1.5-12.0) minimal silanol activity better efficiency than other high pH compatible columns high loading capcity for basic compounds . Metal impurity\<30 ppm; Surface Area: 240m2/g Pore Size: 150
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN DC C8 4.6X150 3U 89402-400 DC831505-0 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty