Would you like to log in again?
TruCool™ Hinged Lid CryoBoxes
Supplier: BioCision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
TruCool®
BCS-209P BCS-217P BCS-207B BCS-221G BCS-206P BCS-207PK BCS-220P BCS-206G BCS-207 BCS-215G BCS-207P BCS-219G BCS-206O BCS-206MC BCS-206PK BCS-207O BCS-206 BCS-209G BCS-206B BCS-217G BCS-219P BCS-215P BCS-221P BCS-207G
89402-720PK 645.47 USD InStock
89402-720 10118-928 89402-722 89402-700 89402-710 10118-926 10118-930 10118-876 10118-874 10118-868 10118-932 10118-878 89427-664 10118-882 10118-880 10118-870 89402-706 89402-716 89402-708 89402-718 89402-702 89402-712 89402-704 89402-714
TruCool™ Hinged Lid CryoBoxes
Boxes Cryoboxes
BioCision® TruCool™ Cryoboxes have a hinged lid that remains attached, ensuring the lid is fitted correctly and markings are in sync with samples
TruCool™ CryoBoxes are equipped with dual indexing systems, including an adjustable removable plastic grid. They feature a 1 through 81 printed grid on the top and inside cover. An A through I, and 1 through 9 column and row indexing around the inner and outer edge of the box. 100-place boxes have a removable 100-place grid and feature an A through J and 1 through 10 column and row indexing around the inner and outer edge of the box.
Designed for repeated freezing and thawing in vapor-phase liquid nitrogen applications. 2-inch boxes are compatible with 1 mL and 2 ml cryovials and microcentrifuge tubes. 3.5-inch boxes are compatible with 3.5 to 5 ml tubes.
Lid is easy to open, even when frozen, and stays attached to the base, minimizing risk of contamination. Boxes feature a white patch on the front for markings or barcodes. Constructed from laminated cardboard, all CryoBoxes are waterproof and durable. Each device includes a plastic grid which can be removed if needed.
Ordering information: Available in single color packs and a multicolor pack of 5 containing one each of blue, green, orange, pink, purple.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty