Would you like to log in again?
TouchNTuff® 93-700, Sterile Nitrile Gloves, Ansell
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
384231 384230 384232 384233
89402-742CS 1203.84 USD InStock
89402-742 89402-744 89402-740 89402-746
TouchNTuff® 93-700, Sterile Nitrile Gloves, Ansell
Gloves
TouchNTuff® Sterile Nitrile Gloves feature an enhanced nitrile formulation for chemical protection, flexibility, comfort, and dexterity.
  • Ambidextrous
  • Excellent chemical protection
  • Low allergenicity
Free of natural rubber latex proteins.
Suggested applications include core pharmaceutical products manufacturing, biotechnology, medical devices, academic research instituitions, and R&D laboratories.
Gamma radiation – sterility assurance level (Sal 10⁻⁶). The 93-700 is an ambidextrous glove with a roll-beaded cuff. Clean sterile pair packs with a pinhole AQL of 1.0. Optimal for critical environment applications.
Certifications: Manufactured in a facility registered to ISO 9001, ISO 13485, ISO14001. Quality control complies with ASTM D 3577, Type II.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty