Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HyFlex® 11-618 Ultralight Gloves, Ansell

Supplier: Ansell

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

HyFlex®
288600 288601 288621 288598 288597 288599
89402-774CS 1232.91 USD InStock
89402-774 89402-776 89402-782 89402-772 89402-780 89402-778
HyFlex® 11-618 Ultralight Gloves, Ansell
Gloves
The next generation glove in comfort, dexterity and flexibility, the HyFlex® 11-618 ultralight gloves maximize touch and precision with a light-duty 18-gauge nylon liner and polyurethane coating that is 20% lighter than other polyurethane/nylon gloves.

  • High abrasion resistance
  • Flexible construction
  • High comfort levels
  • Extended glove life

With a total thickness of 0.8mm, gloves are the thinnest for any delicate handling job while offering outstanding comfort and sensitivity

The polyurethane coating covers only palm and fingers, allowing higher breathability and comfort. In addition, the dark liner and glove mask dirt and reduce glove waste. Designed for a wide range of lightweight applications and good grip in dry or slightly oily environments, gloves are ideal for environments requiring a higher level of touch and precision.

Diverse HyFlex® styles provide the best-in-class comfort, protection and dexterity required for the precise manipulation of small parts, controlled operation of power tools and gear, and direct handling of heavy parts and materials.

Use in the automotive and transportation, building and construction, machinery and equipment, maintenance, and metal fabrication industries. Uses include cosmetics/plastics (also packaging stage); final assembly, airco, electrical engines, wire harness assembly, and adjusters work; finishing works; light assembly applications requiring a high degree of precision; picking and fitting small pieces such as bearings and springs; and white goods and electrical goods.

Certifications: ANSI Level 1 cut protection.

Packaging: Gloves are packaged 12 pairs in a bag; 12 bags in a carton.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty