Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Platform Rocker, Adjustable Tilt, PMR-100, Grant Instruments

Supplier: Grant

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

PMR-100 USA
89402-920EA 3764.54 USD InStock
89402-920
Platform Rocker, Adjustable Tilt, PMR-100, Grant Instruments
Shakers and Mixers Rotating/Rocking Shakers
The sturdy PMR-100 variable speed and adjustable angle rocker is ideally suited for mixing solutions or nutrient media in vessels or bags

Easy to read LCD screen shows actual and set time, speed and set angle. Continuous or timer operation; integral digital timer 1 min to 99 h 59 min, with automatic switch-off, time remaining is displayed, ensures repeatability for time sensitive incubations. Digital speed setting, from 1 to 100, high load capacity up to 5 kg ( 5 litres). Adjustable tilt angle to 9° at lower speeds, fixed at 10° for speeds over 50 giving flexible working. Can be used at 4 to 40 °C (80% relative humidity for temperatures up to 31°C, decreasing linearly to 50% relative humidity at 40°C, non-condensing), in an incubator or cold room.

The unit provides soft or intensive side to side rocking, ensuring optimal sample mixing. Applications include gel destaining, agglutination testing, incubation in immunoassays, homogenisation of extraction media and blots. The reliable motor produces regulated and reproducible rocking throughout the speed range

Certifications: CE Marked.

Ordering information: Supplied with non-slip mat. Manufacturers 2-year warranty.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty