Would you like to log in again?
Multifuge™ X3/X3R Benchtop and Floor Standing Centrifuges, Thermo Scientific
Supplier: Thermo Scientific
75311762 75004531 75011762 75371720 75010817 75366362 75004537 75013839 75011720 75311761 75163839 75163839 75371771 75010817 75311720 75376362 75371761 75377406 75371762 75011761
10324-280EA 22406.76 USD InStock
10324-280 89218-330 10324-262 89402-980 10324-272 89218-334 10324-260 10324-270 10324-266 10324-264 10324-274 CA97040-374 10324-268 CA97040-372 10324-278 97040-374 CA97040-392 97040-392 10033-786 89402-978
Multifuge™ X3/X3R Benchtop and Floor Standing Centrifuges, Thermo Scientific
Centrifuges Benchtop Centrifuges
Process a range of general-purpose applications including cell culture, microplate, bioproduction, blood separation, and microbiology with the Multifuge™ X3 Centrifuge Series
Simple Operation:
- System offers intuitive control panel and digital display.
- Detailed operation information, including display of set and actual parameters, maximize run performance and temperature accuracy for improved sample protection.
- User can easily adjust speed, time, and temperature, even during the run, with a convenient keyboard for direct input.
- Innovative lid locking technology enables one finger downward motion for easiest closing and locking of centrifuge lid, even on tall benches with optional automatic lid opening.
- Double temperature management system displays the air temperature in the chamber as well as the sample temperature, ensuring accurate control of the sample temperature during the run.
Thermo Scientific Auto-Lock™ III Rotor System:
- Secure locking system allows easy push-button installation and exchange of rotors.
- System saves time in switches between applications.
- User has easy access to the chamber, enabling quick cleaning, a healthier working environment, and longer unit life.
Featuring industry-leading throughput and energy efficient operation, this series can process up to 40% more conical tubes in swing-out rotor configuration and 100% more in fixed-angle configuration. A broad range of rotors and adapters, including lightweight Fiberlite™ carbon fiber rotors, handle diverse laboratory applications.
Certifications: UL listed. CE marked. CSA certified. Certified Biosafety.
Ordering information: Accessories are not included.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
SERVICES
Services: Thermo Scientific provides several service options such as Extended Warranty, Extended Warranty plus on-site Annual Preventative Maintenance, and Total Care. To learn more about these services please search "Thermo Centrifuge Technical Service" in the website search field.
In addition, VWRCATALYST™ can provide an array of services such as on-site IQ, OQ, PQ, Customized Protocol Generation, Equipment Calibration, Asset Management, and Certification Services (US only). Please call 888-793-2300 or email VWRCATALYST@vwr.com for further details.
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty