Would you like to log in again?
PDX-LIB Media
Supplier: Hardy Diagnostics
2500325 2500325
89404-952PK 348.36 USD InStock
CA89404-952 89404-952
PDX-LIB Media
Media for Microbiology Dehydrated Media for Microbiology
Media for the detection of Listeria species from the environment, AOAC RI approved, 24 to 30 h result, closed system, no special equipment needed, use with EnviroMAX sampling sponge on a stick 89405-022.
This test is both reliable and inexpensive with presumptive results available for the most common Listeria spp, within 30 h, no sub-culturing or specimen transfers required. Simply collect your sample, inoculate the PDX-LIB media, and incubate. Detect as low as 10 to 50 heat injured Listeria monocytogenes per ml within 24 to 30 h of incubation. Comparable results to USDA methods. The test is AOAC-RI approved. Sampling devices are not included.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty