Would you like to log in again?
Brucella Blood Agar with Hemin, and Vitamin K, Hardy Diagnostics
Supplier: Hardy Diagnostics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
A30 H05 A30
76288-252PK 124.4 USD InStock
76288-252 CA89405-032 89405-032
Brucella Blood Agar with Hemin, and Vitamin K, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the cultivation of anaerobes
A non-inhibitory general growth medium for all gram positive and gram negative anaerobic bacteria. Optimized formula for enhanced growth characteristics of anaerobes.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty