Would you like to log in again?
MUG Supplement, Hardy Diagnostics
Supplier: Hardy Diagnostics
BR071E BR071E
89405-038EA 675.69 USD InStock
89405-038 CA89405-038
MUG Supplement, Hardy Diagnostics
Media for Microbiology Additives and Supplements for Microbiology
A fluorescent agent for the detection of Escherichia coli (contains 4-methylumbelifery-B-D-glucuronide 50 mg).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty