Would you like to log in again?
CulGenex™ Yeast Peptone Dextrose (YPD), Dehydrated Culture Media, Hardy Diagnostics
Supplier: Hardy Diagnostics
C1159 C1159
89405-044PK 565.62 USD InStock
CA89405-044 89405-044
CulGenex™ Yeast Peptone Dextrose (YPD), Dehydrated Culture Media, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CulGenex™ Yeast Peptone Dextrose (YPD) Powders are recommended for the rapid growth of yeasts, particularly Saccharomyces cerevisiae, used for molecular research.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty