Would you like to log in again?
Bacillus cereus Medium, CRITERION™, Hardy Diagnostics
Supplier: Hardy Diagnostics
CRITERION™
C5060 C5062 C5061 C5063 C5060 C5061 C5062 C5063
89405-102EA 2658.12 USD InStock
CA89405-096 CA89405-098 CA89405-100 CA89405-102 89405-102 89405-100 89405-098 89405-096
Bacillus cereus Medium, CRITERION™, Hardy Diagnostics
Media for Microbiology Additives and Supplements for Microbiology
CRITERION™ Bacillus Cereus Agar Base is a selective agar medium used for the isolation and determination of Bacillus Cereus
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Egg Yolk Emulsion, product r450290, and Bacillus Cereus Selective Supplement, Polymyxin B, product #SR099E, are required for this medium. Requires supplements: Egg Yolk Suspension 89407-700, and Bacillus Cereus Selective Supplement Polymyxin B SR099E. Dehydratedculture media with engineered container.
Certifications: BSE Free
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty