Would you like to log in again?
CRITERION™ Bismuth Sulfite Agar
Supplier: Hardy Diagnostics
CRITERION™
C5213 C5212 C5210 C5212 C5211 C5210 C5211 C5213
89405-194EA 378.83 USD InStock
89405-194 CA89405-194 89405-192 CA89405-196 CA89405-198 89405-198 89405-196 CA89405-192
CRITERION™ Bismuth Sulfite Agar
Media for Microbiology Dehydrated Media for Microbiology
Bismuth sulfite agar for the selective isolation of Salmonella spp from clinical and food specimens. Dehydratedculture media with engineered container.
  • Wide mouth opening - reduces dust formation by allowing the easy entry of a scoop
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers - for efficient storage
  • Increased solubility - dissolves quickly
  • Large selection, over 200 formulas available
Certifications: BSE Free.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty