Would you like to log in again?
Columbia CNA Agar Base, CRITERION™, Hardy Diagnostics
Supplier: Hardy Diagnostics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CRITERION™
C5471 C5471 C5472 C5473 C5470 C5472 C5473 C5470
89405-310EA 649.33 USD InStock
CA89405-308 89405-310 CA89405-310 89405-312 CA89405-312 89405-306 89405-308 CA89405-306
Columbia CNA Agar Base, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Columbia CNA Agar Base is intended to be used as a selective basal media to which blood is added for the selective isolation and differentiation of gram-positive cocci.
Dehydratedculture media with engineered container.
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Custom and bulk products available. BSE Free documentation available. Over 30 years of formulation experience
CRITERION™ Columbia CNA Agar Base is intended to be used as a selective basal media to which blood is added for the selective isolation and differentiation of gram-positive cocci.
Certifications: BSE Free
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty