Would you like to log in again?
Kligler Iron Agar, CRITERION™, Hardy Diagnostics
Supplier: Hardy Diagnostics
CRITERION™
C5913 C5913 C5911 C5911 C5912 C5910 C5910 C5912
89405-512EA 61.79 USD InStock
CA89405-518 89405-512 89405-514 CA89405-512 CA89405-514 89405-516 89405-518 CA89405-516
Kligler Iron Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Kligler Iron Agar (KIA) is recommended for use in differentiating certain members of the Enterobacteriaceae by demonstrating hydrogen sulfide production and the fermentation of dextrose and lactose
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty