Would you like to log in again?
Lactose, (Milk Sugar), CRITERION™, Hardy Diagnostics
Supplier: Hardy Diagnostics
C5953 C5952 C5951 C5950 C5953 C5950 C5952 C5951
89405-540EA 362.44 USD InStock
89405-540 89405-542 CA89405-540 CA89405-542 89405-536 CA89405-536 89405-538 CA89405-538
Lactose, (Milk Sugar), CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Lactose (Milk Sugar) is chemically pure D-lactose monohydrate
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
It is added to culture media as a source of energy for bacteria and for the determination of fermentation reactions. Dehydratedculture media with engineered container.
Certifications: BSE Free
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty