Would you like to log in again?
MacConkey Agar with MUG, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
CRITERION™
C6110 C6111 C6113 C6110 C6111 C6112 C6113 C6112
89405-618EA 2623.24 USD InStock
CA89405-616 89405-618 CA89405-618 89405-612 CA89405-612 89405-614 CA89405-614 89405-616
MacConkey Agar with MUG, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ MacConkey Agar with MUG is used for the presumptive identification of Escherichia coli. Dehydratedculture media with engineered container.
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Certifications: BSE Free
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty