Would you like to log in again?
MacConkey Agar with Sorbitol, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
CRITERION™
C6171 C6170 C6171 C6172 C6170 C6173 C6172 C6173
89405-650EA 1856.13 USD InStock
89405-650 CA89405-650 89405-644 89405-646 CA89405-644 CA89405-646 89405-648 CA89405-648
MacConkey Agar with Sorbitol, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ MacConkey Agar with Sorbitol is to be used as a selective and differential medium for the detection of enterohemorrhagic Escherichia coli 0157:H7. Dehydratedculture media with engineered container.
· Wide mouth opening - reduces dust formation by allowing the easy entry of a scoop
· Convenient handgrip - 'no slip' finger holds
· Stackable containers - for efficient storage
· Increased solubility - dissolves quickly
· Large selection, over 200 formulas available
· Custom and bulk products available - to your specifications
· Tested for optimal performance
· BSE Free (contact for specifics)
· Over 30 years of formulation experience
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Custom and bulk products available. BSE Free documentation available. Over 30 years of formulation experience
CRITERION™ MacConkey Agar with Sorbitol is to be used as a selective and differential medium for the detection of enterohemorrhagic Escherichia coli 0157:H7
Dehydratedculture media with engineered container.
Certifications: BSE Free
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty