Would you like to log in again?
Malt Extract Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
CRITERION™
C6202 C6201 C6201 C6200 C6202 C6203 C6203 C6200
89405-652EA 57.3 USD InStock
89405-652 89405-654 CA89405-652 CA89405-654 89405-656 CA89405-656 89405-658 CA89405-658
Malt Extract Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Malt Extract Agar for the isolation, cultivation and maintenance of yeasts and molds. Dehydratedculture media with engineered container.
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Custom and bulk products available. BSE Free documentation available. Over 30 years of formulation experience
CRITERION™ Malt Extract Agar for the isolation, cultivation and maintenance of yeasts and molds
Dehydratedculture media with engineered container.
Certifications: BSE Free
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty