Would you like to log in again?
Mycobacteria 7H11 Agar Base, CRITERION™, Hardy Diagnostics
Supplier: Hardy Diagnostics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CRITERION™
C6292 C6290 C6293 C6292 C6290 C6291 C6293 C6291
89405-706EA 1555.81 USD InStock
CA89405-704 89405-706 CA89405-706 89405-700 CA89405-700 89405-702 89405-704 CA89405-702
Mycobacteria 7H11 Agar Base, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Mycobacteria 7H11 Agar Base for the cultivation of Mycobacterium spp
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
OADC Supplement, product 89407-732 and Glycerol, product 89404-938, are required for this medium. Dehydratedculture media with engineered container.
Certifications: BSE Free
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty