Would you like to log in again?
Motility Med S Base, CRITERION™, Hardy Diagnostics
Supplier: Hardy Diagnostics
C6390 C6393 C6390 C6392 C6392 C6393 C6391 C6391
89405-752EA 89 USD InStock
89405-752 CA89405-752 89405-754 89405-756 CA89405-754 89405-758 CA89405-756 CA89405-758
Motility Med S Base, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Motility Med S Base for the detection of bacterial motility
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
TTC Solution (1%), product 89404-940, is required for this medium. Dehydratedculture media with engineered container.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty