Would you like to log in again?
Nitrate Broth, CRITERION™, Hardy Diagnostics
Supplier: Hardy Diagnostics
C6451 C6452 C6453 C6452 C6450 C6453 C6451 C6450
89405-780EA 499.77 USD InStock
89405-780 89405-782 CA89405-780 CA89405-782 89405-776 CA89405-776 89405-778 CA89405-778
Nitrate Broth, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Nitrate Broth is recommended for differentiating microorganisms based upon nitrate reduction
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty