Would you like to log in again?
Nutrient Gelatin, CRITERION™, Hardy Diagnostics
Supplier: Hardy Diagnostics
C6482 C6481 C6480 C6481
89405-800EA 129.08 USD InStock
89405-800 89405-802 89405-804 CA89405-802
Nutrient Gelatin, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Nutrient Gelatin is used to aid in classification of the Enterobacteriaceae by determining an organism's ability to liquefy gelatin. Dehydratedculture media with engineered container.
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
· Wide mouth opening - reduces dust formation by allowing the easy entry of a scoop
· Convenient handgrip - 'no slip' finger holds
· Stackable containers - for efficient storage
· Increase solubility - dissolves quickly
· Custom and bulk products available to your specifications
· Tested for optimal performance
· BSE Free (contact for specifics)
· Over 30 years of formulation experience
Custom and bulk products available. BSE Free documentation available. Over 30 years of formulation experience
Custom and bulk products available
BSE Free documentation available. Over 30 years of formulation experience
Certifications: BSE Free
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty