Would you like to log in again?
CRITERION™ Peptone Water
Supplier: Hardy Diagnostics
CRITERION™
C6571 C6573 C6570 C6573 C6570 C6571 C6572 C6572
89405-870EA 357.66 USD InStock
89405-870 89405-872 CA89405-870 CA89405-872 89405-866 89405-868 CA89405-866 CA89405-868
CRITERION™ Peptone Water
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ peptone water is used for the cultivation of nonfastidious organisms, for performing the indole test, and for studying fermentation patterns.
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty