Would you like to log in again?
Rappaport-Vassiliadis R10 Broth, Hardy Diagnostics
Supplier: Hardy Diagnostics
C6731 C6731
89405-956EA 260.94 USD InStock
89405-956 CA89405-956
Rappaport-Vassiliadis R10 Broth, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Rappaport-Vassiliadis R10 Broth is primarily for use in the selective enrichment and isolation of <italic>Salmonella spp
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
from food and environmental specimens.Dehydratedculture media with engineered container.</italic>
Certifications: BSE Free
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty