Would you like to log in again?
Rappaport-Vassiliadis (MSRV) Medium Base, Semisolid Modification, CRITERION™, Hardy Diagnostics
Supplier: Hardy Diagnostics
C6742 C6740 C6743 C6742 C6741 C6743 C6741 C6740
89405-960EA 296.71 USD InStock
CA89405-958 89405-960 89405-962 CA89405-960 CA89405-962 89405-964 CA89405-964 89405-958
Rappaport-Vassiliadis (MSRV) Medium Base, Semisolid Modification, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Rappaport-Vassiliadis (MSRV) Medium Base, along with novobiocin, is used in the detection and isolation of motile Salmonella spp. in feces and food products
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Sodium Novobiocin, product #SR181E, is required for this medium. Dehydratedculture media with engineered container.
Certifications: BSE Free
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty