Would you like to log in again?
Sabdex Agar with Chloramphenicol, CRITERION™, Hardy Diagnostics
Supplier: Hardy Diagnostics
CRITERION™
C6782 C6783 C6780 C6780 C6781 C6782 C6781 C6783
89405-980EA 931.57 USD InStock
89405-980 CA89405-980 89405-974 CA89405-974 89405-976 89405-978 CA89405-976 CA89405-978
Sabdex Agar with Chloramphenicol, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Sabdex Agar with Chloramphenicol is recommended as a selective agar medium for the isolation, identification and cultivation of fungi
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty