Would you like to log in again?
Sabdex Agar, CRITERION™, Hardy Diagnostics
Supplier: Hardy Diagnostics
CRITERION™
C6811 C6810 C6811 C6810 C6813 C6813 C6812 C6812
89406-004EA 1468.99 USD InStock
CA89406-004 89406-004 CA89406-002 CA89406-000 89406-002 89406-000 89405-998 CA89405-998
Sabdex Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Sabouraud Dextrose Agar (Sabdex) is recommended for the isolation, cultivation, and maintenance of non-pathogenic and pathogenic species of fungi and yeasts
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty