Would you like to log in again?
Starch Agar, CRITERION™, Hardy Diagnostics
Supplier: Hardy Diagnostics
CRITERION™
C7010 C7012 C7013 C7012 C7010 C7011 C7011 C7013
89406-092EA 2055.08 USD InStock
CA89406-092 CA89406-090 89406-092 89406-090 CA89406-088 CA89406-086 89406-088 89406-086
Starch Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Starch Agar is used for cultivating microorganisms being tested for starch hydrolysis
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty