Would you like to log in again?
CRITERION™ Sucrose
Supplier: Hardy Diagnostics
CRITERION™
C7021 C7020 C7022 C7023 C7021 C7022 C7023 C7020
89406-094EA 51.89 USD InStock
89406-094 CA89406-100 CA89406-098 89406-100 CA89406-096 89406-098 CA89406-094 89406-096
CRITERION™ Sucrose
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Sucrose is added to culture media for use in preparing microbiological culture media.
  • Wide mouth opening
  • Reduces dust and allows easy scooping
  • Convenient handgrip
  • 'No slip' finger holds
  • Stackable containers
  • High solubility; dissolves quickly
  • Custom and bulk products available
  • Unparalleled performance - tested for optimal growth
For fermentation tests it is usually added at a final concentration of 0.5 to 1.0%. Dehydratedculture media with engineered container.
These culture media is formulated to meet or exceed the highest standards.
Application: Breweries, wineries and distilleries, cannabis QA testing, clinical, cosmetics, environmental, food safety, compounding pharmacy, pharmaceutical manufacturing
Certifications: TSE/BSE free.
Delivery information: 500 g - wide mouth, easy grip bottle, comes with a free scoop, 2 and 10 kg bucket - with convenient screw lid.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty