Would you like to log in again?
Triple Sugar Iron Agar, CRITERION™, Hardy Diagnostics
Supplier: Hardy Diagnostics
CRITERION™
C7112 C7113 C7110 C7113 C7112 C7111 C7111 C7110
89406-156EA 1069.52 USD InStock
CA89406-156 89406-156 CA89406-154 89406-154 CA89406-152 CA89406-150 89406-152 89406-150
Triple Sugar Iron Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Triple Sugar Iron (TSI) Agar is recommended for use in the differentiation of Enterobacteriaceae by their ability to ferment glucose, lactose, and sucrose, and produce hydrogen sulfide.
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Certifications: BSE Free
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty