Would you like to log in again?
Tryptic Soy Agar (TSA), CRITERION™, Hardy Diagnostics
Supplier: Hardy Diagnostics
CRITERION™
C7120 C7124 C7123 C7120 C7121 C7122 C7124 C7121 C7122 C7123
89406-160EA 141.38 USD InStock
89406-160 CA89406-158 89406-158 CA89406-166 89406-166 CA89406-164 89406-164 CA89406-162 89406-162 CA89406-160
Tryptic Soy Agar (TSA), CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Tryptic Soy Agar is recommended for use as a general growth medium for the isolation and cultivation of microorganisms
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
This medium is also recommended for use in the cultivation, storage, and transportation of pure cultures of bacteria.
Certifications: BSE Free
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty