Would you like to log in again?
Tryptose Phosphate Broth, CRITERION™, Hardy Diagnostics
Supplier: Hardy Diagnostics
C7211 C7211 C7213
89406-208EA 2447.49 USD InStock
CA89406-206 89406-208 89406-206
Tryptose Phosphate Broth, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Mycobiotic Agar is recommended for use in the selective isolation and recovery of pathogenic fungi and dermatophytes from clinical specimens; especially hair, skin, and nails.
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Certifications: BSE Free
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty