Would you like to log in again?
Violet Red Bile Agar with MUG, CRITERION™, Hardy Diagnostics
Supplier: Hardy Diagnostics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CRITERION™
C7252 C7250 C7253 C7250 C7253 C7251 C7252 C7251
89406-228EA 173.4 USD InStock
CA89406-228 CA89406-226 89406-228 89406-226 CA89406-232 89406-232 CA89406-230 89406-230
Violet Red Bile Agar with MUG, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Violet Red Bile Agar with MUG is primarily for use in the detection of Escherichia coli and total coliforms in food and dairy products.
CRITERION™ Dehydrated Culture Media and container.
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Custom and bulk products available. BSE Free documentation available. Over 30 years of formulation experience
CRITERION™ Violet Red Bile Agar with MUG is primarily for use in the detection of Escherichia coli and total coliforms in food and dairy products.
Certifications: BSE Free
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty