Would you like to log in again?
Letheen Broth, Modified, CRITERION™, Hardy Diagnostics
Supplier: Hardy Diagnostics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
C7643 C7640 C7643 C7642 C7640 C7641 C7641 C7642
89406-466EA 2149.97 USD InStock
CA89406-466 89406-466 CA89406-464 CA89406-462 89406-464 CA89406-460 89406-462 89406-460
Letheen Broth, Modified, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Letheen Broth, Modified, for the determination of antimicrobial activity of quaternary ammonium compounds
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty