Would you like to log in again?
m PA-C Agar, CRITERION™, Hardy Diagnostics
Supplier: Hardy Diagnostics
CRITERION™
C7961 C7960 C7961 C7963 C7963 C7962 C7960 C7962
89406-604EA 92 USD InStock
89406-604 CA89406-610 89406-610 CA89406-608 CA89406-606 89406-608 89406-606 CA89406-604
m PA-C Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ m PA-C Agar is a modified formulation for the cultivation and enumeration of Pseudomonas aeruginosa in water by membrane filtration
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty