Would you like to log in again?
CRITERION™ Tryptose, Hardy Diagnostics
Supplier: Hardy Diagnostics
C8073 C8071 C8071 C8070 C8073
89406-668EA 220.59 USD InStock
CA89406-668 89406-668 CA89406-666 CA89406-672 89406-672
CRITERION™ Tryptose, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Tryptose is primarily used for preparing microbiological culture media and when non-animal source nutrients are needed in the cultivation of fastidious and non-fastidious organsims
  • Non-mammalian
  • Wide mouth opening
  • Reduces dust and allows easy scooping
  • Convenient handgrip
  • 'No slip' finger holds
  • Stackable containers
  • High solubility; dissolves quickly
  • Custom and bulk products available
Dehydrated culture media with engineered container. Custom and bulk products available. BSE Free documentation available. Over 30 years of formulation experience.
Certifications: TSE/BSE Free
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty