Would you like to log in again?
Modified Plate Count Agar, CRITERION™, Hardy Diagnostics
Supplier: Hardy Diagnostics
CRITERION™
C8193 C8192 C8190 C8192 C8191 C8191 C8190 C8193
89406-722EA 2251.28 USD InStock
CA89406-722 89406-722 CA89406-720 89406-720 CA89406-718 CA89406-716 89406-718 89406-716
Modified Plate Count Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
CRITERION™ Modified Plate Count Agar, (Plate Count Agar with Bromcresol Purple and Tween® 80), is recommended for the cultivation and differentiation of Lactobacilli
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty