Would you like to log in again?
CulGenex™ Antibiotic and Supplements
Supplier: Hardy Diagnostics
CG19 CG17 CG18 CG21 CG21 CG17 CG16 CG20 CG20 CG18 CG19
89406-888PK 81.28 USD InStock
CA89406-888 CA89406-910 CA89406-900 89406-888 CA89406-886 89406-910 89406-900 CA89406-916 CA89406-890 89406-916 89406-890
CulGenex™ Antibiotic and Supplements
Media for Microbiology Additives and Supplements for Microbiology
CulGenex™ Antibiotic and Chromogenic Supplements are recommended for use in the selective and/or differential growth of recombinant strains of Escherichia coli used for genetic research.
Antibiotics such as ampicillin, kanamycin, and carbenicillin are typically used as selective agents to isolate strains that have taken up genes (e.g. plasmids) coupled to a gene encoding for resistance. X-GAL, used in conjunction with IPTG, is intended for the colorimetric detection of beta-galactosidase activity in recombinant strains.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty