Would you like to log in again?
Pseudomonas Isolation Agar, 15×60 mm plate, Hardy Diagnostics
Supplier: Hardy Diagnostics
G145 G145
89407-166PK 86.05 USD InStock
CA89407-166 89407-166
Pseudomonas Isolation Agar, 15×60 mm plate, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
For the selective isolation of Pseudomonas aeruginosa
Pseudomonas Agar P is recommended for enhancement of pyocyanin production by Pseudomonas aeruginosa.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty