Would you like to log in again?
HE (Hektoen Enteric) Agar, Hardy Diagnostics
Supplier: Hardy Diagnostics
G63 G63
89407-338PK 26.53 USD InStock
89407-338 CA89407-338
HE (Hektoen Enteric) Agar, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Hektoen Enteric Agar (HE) is for the selective isolation and differentiation of pathogenic Gram negative enteric bacteria, such as the non-lactose fermenting <italic>Salmonella</italic> species and <italic>Shigella</italic> species Normal stool flora, such as <italic>E.coli</italic>, will be partially inhibited.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty