Would you like to log in again?
Lok-Tight™ Contact Plates, Hardy Diagnostics
Supplier: Hardy Diagnostics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
P98 P32 P80 P46 P98 P80 P93 P33 P36 P42 P51 P09 P15 P99 P99 P09 P27 P34 P93 P51 P15 P36 P33 P32 P47 P42 P34
89407-878PK 56.43 USD InStock
CA89407-842 89407-878 89407-714 89407-510 76308-442 89407-697 CA89407-860 CA89407-728 CA89407-708 10153-328 CA89407-946 89407-816 CA89408-026 CA89407-396 89407-860 89407-482 CA89407-510 89407-842 89407-708 CA89407-816 CA89407-878 89407-946 CA89407-714 89407-728 89408-026 CA89407-482 89407-396
Lok-Tight™ Contact Plates, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
An added measure to ensure sterility in your environmental testing program.
  • Friction lid design keeps lids in place
  • Lok-Tight™ optional locking feature means lids stay secured during sampling and transport
  • Each plate has a grid molded into the bottom of the plate to aid in the counting of microorganisms
Hardy Diagnostics contact plates are recommended for use in the cultivation of microorganisms from environmental surfaces.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty