Would you like to log in again?
Buffered Sodium Chloride Peptone EP, USP, Hardy Diagnostics
Supplier: Hardy Diagnostics
U301
89407-428PK 424.98 USD InStock
89407-428
Buffered Sodium Chloride Peptone EP, USP, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Use as rinse fluid for membrane filtration methods.
Use as rinse fluid for membrane filtration methods.
Ordering information: Minimum order of 5 packs of 10.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty