Would you like to log in again?
Buffered Peptone Water, Hardy Diagnostics
Supplier: Hardy Diagnostics
U142 U143
89407-426PK 285.66 USD InStock
89407-426 89407-434
Buffered Peptone Water, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
A preenrichment used for increasing the recovery of injured Salmonella spp
A preenrichment used for increasing the recovery of injured Salmonella spp. from food and other specimens prior to selective enrichment and isolation.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty