Would you like to log in again?
Cetrimide Selective/MacConkey/Vogel and Johnson Agar triplate
Supplier: Hardy Diagnostics
J314
89407-452PK 70.1 USD InStock
89407-452
Cetrimide Selective/MacConkey/Vogel and Johnson Agar triplate
Media for Microbiology Prepared Media for Microbiology
For isolation and differential of gram-negative bacteria.
Section III: Vogel and Johnson Agar; For selective isolation of coagulase-positive and mannitol-positive S. aureus.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty