Would you like to log in again?
CoaguStaph™, Rabbit Coagulase Plasma with EDTA, Hardy Diagnostics
Supplier: Hardy Diagnostics
Z020 Z020 Z003 Z003
89407-458EA 145.17 USD InStock
89407-458 CA89407-458 CA89407-460 89407-460
CoaguStaph™, Rabbit Coagulase Plasma with EDTA, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Lyophilized rabbit plasma, for the identification of Staphylococcus aureus</italic by the coagulase test
EDTA helps eliminate false positives, not recommended for germ tubes, packaged in a vial with a rubber septum and aluminum crimp sea
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty