Would you like to log in again?
E. coliPRO™ O157 Latex Kit, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
PL070HD PL070HD
89407-502EA 312 USD InStock
89407-502 CA89407-502
E. coliPRO™ O157 Latex Kit, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
A rapid latex agglutination test kit for the identification of the E.coli O157 serotype.
So sensitive that it requires only one single colony. Uses proprietary antibodies that eliminate the possibility of cross-reactions with Escherichia hermanii and other sorbitol-negative Escherichia species. Contains both positive and negative controls. Includes reaction cards and mixing sticks. Rapid results in just 2 minutes. 2 year shelf life.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty