Would you like to log in again?
EnviroTrans™, Saline 0.85%, 0.5 ml Swab, Hardy Diagnostics
Supplier: Hardy Diagnostics
SRK45 SRK55 SRK35 SRK15 SRK55 SRK30 SRK35 SRK30 SRK05 SRK20 SRK25 SRK20 SRK25 SRK05 SRK45
89407-766PK 76.4 USD InStock
89407-766 CA89407-764 CA89407-512 89407-912 CA89407-672 89407-764 89407-512 CA89407-916 89407-914 CA89407-912 CA89407-914 89407-916 CA89407-470 89407-672 89407-470
EnviroTrans™, Saline 0.85%, 0.5 ml Swab, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
A ready-to-use swab & tube pre-filled with your choice of five solutions to fit your specific needs! Collect and transport your environmental sample with efficiency and ease
Remove the cap attached to the swab. Collect sample by swabbing the surface, recap, and send to the lab. A 15 x 75 mm polypropylene tube pre-filled with 5mls of Letheen Broth. A dacron swab is affixed to a screw cap for easy handling. The entire unit is terminally sterilized. Excellent for detection or enumeration of microorganisms from environmental surfaces or equipment. Ideal for your HACCP program!.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty