Would you like to log in again?
Fluid K, USP, Hardy Diagnostics
Supplier: Hardy Diagnostics
U206
89407-548PK 330.5 USD InStock
89407-548
Fluid K, USP, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
A diluting and rinsing fluid for the membrane filtration technique.
  • Certified for pH, sterility and growth promotion
Hardy Diagnostics' sterility media and rinse solutions are manufactured in an ISO 13485 environmentally controlled production center. Each lot is certified for pH, sterility and growth promotion using ATCC® strains specified by the USP.
Certifications: Certified for pH, sterility and growth promotion.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty