Would you like to log in again?
Fraser Broth, Modified, Hardy Diagnostics
Supplier: Hardy Diagnostics
K98 K98
89407-598PK 157.91 USD InStock
89407-598 CA89407-598
Fraser Broth, Modified, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the selective enrichment of Listeria species (without ferric ammonium citrate).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty