Would you like to log in again?
Letheen Broth, Hardy Diagnostics
Supplier: Hardy Diagnostics
K207 K106 K207 K118 K118 K106
89407-674PK 107.12 USD InStock
CA89407-666 89407-674 89407-664 CA89407-674 CA89407-664 89407-666
Letheen Broth, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For use in determining bactericidal activity of quaternary ammonium compounds.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty